Learn More
Cole-Parmer Environmental Express BPC1506 Pre-Cleaned Oblong Wide-Mouth Bottle, HDPE, Level 1, 125 mL; 500/CS
Catalog No. NC2103780 Shop All Cole-Parmer Products
Encompass
Store and retrieve samples with easeOblong shape minimizes dead space between bottles when storedKeep samples safe with leakproof capLevel 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratoryLevel 3 containers meet EPA guidelines as well, but do not ship with a certificate
This item is not returnable. View return policy
Catalog No. NC2103780 Supplier Cole-Parmer Supplier No. 3520606
Store and retrieve samples with easeOblong shape minimizes dead space between bottles when storedKeep samples safe with leakproof capLevel 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratoryLevel 3 containers meet EPA guidelines as well, but do not ship with a certificate