Learn More
Abcam Pre-coated 384 well Microplate SimpleStep ELISA, 1UNIT
Supplier:  Abcam AB2033591UNIT
Encompass_Preferred
384-Well SimpleStep ELISA Microplate is an alternative to the 96-well microplate included with all SimpleStep ELISA kits - Combine the reagents from 2 x 96-well SimpleStep ELISA kit to increase your throughput to 384-well - Protocol and recommendations available.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1933679