Learn More
Analytical Sales & Services Pre-Column Pre-Heater
Catalog No. 5000804888
Encompass
Advantage Pre-Column Pre-Heater 0.010" I.D. x 30cm Stainless Steel 15.210 µL Internal Volume With (1) UHPLC Fitting
Qty/Pkg: EACH
Catalog No. 50-008-04888 Supplier Analytical Sales & Services Supplier No. ADV9000UHP
Advantage Pre-Column Pre-Heater 0.010" I.D. x 30cm Stainless Steel 15.210 µL Internal Volume With (1) UHPLC Fitting
Qty/Pkg: EACH