Learn More
THORLABS INC Precision Cover Glasses, #1.5H Thickness, ¿2 mm, Pack of 100
Catalog No. NC1851409
Encompass
Precision Cover Glasses, #1.5H Thickness, ¿2 mm, Pack of 100
Catalog No. NC1851409 Supplier THORLABS INC Supplier No. CG15NH1
Precision Cover Glasses, #1.5H Thickness, ¿2 mm, Pack of 100