Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pre-ELM-11 98.0+%, TCI America™
SDP

Catalog No. C24095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24095G Supplier TCI America Supplier No. C24095G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 854623-98-6
Molecular Formula C20H20B2CuF8N4O2
Molecular Weight (g/mol) 585.559
InChI Key OLBITSHZKINIHL-UHFFFAOYSA-N
Synonym [Cu(bpy)(BF4)2(H2O)2]bpy, Precursor-ELM-11
PubChem CID 11592343
IUPAC Name copper;2-pyridin-2-ylpyridine;ditetrafluoroborate;dihydrate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cu+2]

Specifications

Color Blue-White
UN Number 1759
Quantity 5 g
Formula Weight 585.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material pre-ELM-11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.