Learn More
ACROBiosystems pre-Gc protein 50ug
Supplier:  ACROBiosystems PRNV52H450UG
Encompass_Preferred
Hantaan virus (strain 76-118) (Korean hemorrhagic fever virus) pre-Gc protein His Tag (PRN-V52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 649 - Asn 1105 (Accession # P08668).
Catalog No. 50-253-4283