Learn More
Anatrace NG310-CH210 - 10:1 LMNG/CHS Pre-Made Solution
Catalog No. NC1495364
Encompass
Lauryl Maltose Neopentyl Glycol (NG310) and Cholesteryl Hemisuccinate (CH210) Solution • LMNG/CHS Solution • Lauryl Maltose Neopentyl Glycol (50 mg/ml):CHS-TRIS (5 mg/ml) SolutionCAS Number: LMNG: [1257852-96-2] / CHS: [102601-49-0]Formula Weight: LMNG: 1005.19 / CHS: 607.9
Catalog No. NC1495364 Supplier Anatrace Supplier No. NG310CH210 50 ML
Lauryl Maltose Neopentyl Glycol (NG310) and Cholesteryl Hemisuccinate (CH210) Solution • LMNG/CHS Solution • Lauryl Maltose Neopentyl Glycol (50 mg/ml):CHS-TRIS (5 mg/ml) SolutionCAS Number: LMNG: [1257852-96-2] / CHS: [102601-49-0]Formula Weight: LMNG: 1005.19 / CHS: 607.9