Learn More
Crescent Chemical Co Inc PRE-MIXED MODIFIER 100ML
SDP
Catalog No. 5079732
Encompass_Preferred
Pre-Mixed Modifier 0.75% Pd/0.05% Mg(NO3)2 2% in HNO3 CAS Size - 250 mL Catalog Number - PRE-003
Catalog No. 50-797-32 Supplier Crescent Chemical Co Inc Supplier No. CTPRE003
Pre-Mixed Modifier 0.75% Pd/0.05% Mg(NO3)2 2% in HNO3 CAS Size - 250 mL Catalog Number - PRE-003