Learn More
World Precision Instrument Pre-polarizer for Nitric Oxide Sensors
SDP
Catalog No. 502521057
Encompass
Keep extra NO sen­sors ready to use
Achieve a stable background current quickly. This small battery-powered device applies a potential to the NO electrode equivalent to the potential applied by the ISO-NO meter. Consequently, a sensor which has been connected to the activator may be transferred to the meter for immediate use. For use with all WPI NO electrodes.
Catalog No. 50-252-1057 Supplier World Precision Instrument Supplier No. NSA3
Keep extra NO sen­sors ready to use
Achieve a stable background current quickly. This small battery-powered device applies a potential to the NO electrode equivalent to the potential applied by the ISO-NO meter. Consequently, a sensor which has been connected to the activator may be transferred to the meter for immediate use. For use with all WPI NO electrodes.