Learn More
Miles Scientific PRE-SCRAPED 2000UM ROTOR
SDP
Supplier:  Miles Scientific R02205P
Encompass_Preferred
Rotor; Pre-scraped; Uniform layer; High performance silica gel GF 2000um; For CycloGraph and Chromatotron; UV254 Indicator; Precision cut glass
Catalog No. NC9364402