Learn More
Seekonk Manufacturing Co Inc PRE SET SLIP TORQUE WRENCH
SDP
Supplier:  Seekonk Manufacturing Co Inc LTL
Encompass_Preferred
l handle pre set slip type torque wrench 3/8"dr.5-60in lbs.modes:torque righ-lock left.must specify torque setting needed
Catalog No. NC2180116