Learn More
Analytical Sales & Services Pre-Slit Cap Mat for 350µL Select-A-Vial™ Systems
Catalog No. 5000806403
Encompass
Beige Rubber Cap Mat\n• Made of thermoplastic elastomer\n• Pre-slit\n• Fits 350µL Select-A-Vial™ System with Conical Inserts\n• Can be used with 8 x 30mm Crimp Vials\n\nQty/Pkg: 5
Catalog No. 50-008-06403 Supplier Analytical Sales & Services Supplier No. 35075
Beige Rubber Cap Mat\n• Made of thermoplastic elastomer\n• Pre-slit\n• Fits 350µL Select-A-Vial™ System with Conical Inserts\n• Can be used with 8 x 30mm Crimp Vials\n\nQty/Pkg: 5