Learn More
BD FACS™ Pre-Sort Buffer, 250 mL
Catalog No. BDB563503 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
BDB563503 250 mL
1 options
Catalog No. BDB563503 Supplier BD FACS™ Supplier No. 563503
Add to Cart
Add to Cart
For research use only
BD FACS™ Pre-Sort Buffer was designed to aid in preparing and maintaining single cell suspensions prepared from lymphoid tissues, bone marrow, peripheral blood, cultured cells (including pluripotent stem cells) or other sources. Additives may be necessary for sensitive cell types requiring specific nutrients. The Pre-Sort Buffer contains no phenol red and is an optically clear buffer to help minimize background. The buffer has been formulated to minimize cell clumping which facilitates consistent drop formation on fluorescence activated cell sorters. The buffer has minimal calcium and magnesium to minimize cell aggregation as these cations are necessary cofactors for many cell adhesion molecules. The Pre-Sort Buffer contains an FBS-based protein to help maintain cells in a viable state during cell sorting applications but the buffer does not contain EDTA, which can have deleterious effects on cells. It is useful for the dilution and application of fluorescent reagents as well as for the suspension, washing, and storage of cells destined for florescence activated cell sorting (FACS) or flow cytometric analysis.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Research Use Only
For Use With (Equipment) Flow Cytometer
Quantity 250 mL