Learn More
Keystone Cleanroom Products Pre-Sterilized Media Plate Transport and Incubation Bags

Description
- Ready to Use
- Validated Sterile
- Tyvek / Mylar packaging utilizes hang hooks on each corner to facilitate use in isolators
- Tyvek packaging is compatible with VHP decontamination cycles
- Manufactured and packaged in an ISO certified cleanroom
- Self-sealing Tyvek Peel pouches hold the media plates that are collected inside the isolator
- These Tyvek pouches are transferred to the QC Lab
- The media plates do not need to be removed from the pouches for incubation
- Certificates of Sterility and Conformance with every shipment
- Custom packaging and configurations are available
The system provides complete protection from the time the plates are being removed from inside the isolator all the way through incubation.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | CoC and Certificate of Sterility |
| Closure Type | Self-seal |
| Color | Clear, White |
| Disposable | Single-use |
| For Use With (Application) | For the movement and protection of environmental monitoring media plates from collection through incubation |
| Material | Reinforced Multicoated Plastic |
| Sterility | Sterile |
| Latex Free | Yes |
| No. per Case | 250 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.