Learn More
Keystone Cleanroom Products Pre-Sterilized Sharpie, Fine Point
SDP
Catalog No. 17100849
Change view
Click to view available options
Color:
Black
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-849 Blue
17-100-848 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-100-849 Supplier Keystone Cleanroom Products Supplier No. S10FMARKBLUE
Add to Cart
Add to Cart
Ready-to-use sterile permanent ink Sharpie markers eliminate the risks associated with surface sanitization procedures.
  • Gamma Irradiated Sterile to a 10-6 SAL
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Material Plastic
Ruled No
Color Blue
Cover Color Blue
Autoclavable Not Autoclavable
Certifications/Compliance Certificate of Sterility
Disposable Limited-use
Packaging 10 Pk.
Product Type Sterile Marker
Sterility Sterile
Tip Style Fine Point