Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Cleanroom Products Pre-Sterilized Whirl-Pak Bags
SDP

Catalog No. 19042389
Change view
Click to view available options
No. per Pack:
20 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-042-389 20 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-042-389 Supplier Keystone Cleanroom Products Supplier No. S20319DPST10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pre-sterilized bags designed to reduce the risks of false positives of media plates

  • Designed to reduce the risks of false positives of media plates
  • This system provides complete protection from the time the plates are removed from the isolator all the way through incubation
  • The clear film material offers excellent clarity combined with strength and durability.
  • These bags feature the maximum protection against leaks with the continuous perimeter seal
  • Ready to use, double packed in easy to open Chevron-Peel DuPont™ Tyvek™ pouches, with an extra, sealed poly bag serving as a third sterile packaging layer
  • The inner and outer Tyvek™ iron Peel pouches feature hang holes that can be used to suspend the package inside the isolator for VHP decontamination
  • Validated sterile to a 10-6 SAL
  • Each Tyvek peel pouch package is labeled with an expiration date and a Lot number for complete traceability

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Certificate of Sterility
Color Clear
Disposable Single-use
For Use With (Application) Transport EM Media Plates
Material LDPE
Sterility Sterile
Latex Free Yes
No. per Pack 20 Pk.
Sterilization Method Gamma
Thickness (English) 3 mil
Width (English) 7.5 in.
Product Type Reclosable Bag
Length (Metric) 38.1 cm
Width (Metric) 19.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.