Learn More
Vector Biolabs eGFP Adenovirus, 200ul
Catalog No. 5000333133
Encompass_Preferred
This recombinant human adenovirus type 5 expresses enhanced green fluorescent protein under the control of a CMV promoter. Green fluorescent protein (GFP) is a protein originally isolated from the jellyfish that fluoresces green when exposed to blue light. Enhanced GFP (eGFP) is a GFP mutant with improved fluorescence and stability. Recombinant eGFP adenovirus serves as a control for other recombinant adenoviruses. Since GFP is easily visualized under fluorescence microscopy, this adenovirus can also be used to determine the transduction efficiency and to optimize viral infection condition in a specific cell type.
Catalog No. 50-003-33133 Supplier Vector Biolabs Supplier No. 1060
This recombinant human adenovirus type 5 expresses enhanced green fluorescent protein under the control of a CMV promoter. Green fluorescent protein (GFP) is a protein originally isolated from the jellyfish that fluoresces green when exposed to blue light. Enhanced GFP (eGFP) is a GFP mutant with improved fluorescence and stability. Recombinant eGFP adenovirus serves as a control for other recombinant adenoviruses. Since GFP is easily visualized under fluorescence microscopy, this adenovirus can also be used to determine the transduction efficiency and to optimize viral infection condition in a specific cell type.