Learn More
Vector Biolabs Signal transducer/activator of transcription 5B Adenovirus (Ad-STAT5b)
Supplier:  Vector Biolabs 1467
Encompass_Preferred
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression.
Catalog No. 50-003-33093