Learn More
Chemglass Life Sciences Pre-Trap Only, Airfree, Schlenk
SDP
Catalog No. AF045402
Encompass_Preferred
Pre-Trap Only, Airfree, Schlenk. Pre-trap component of 4-Port Vacuum Manifold Right Handed Trap Assembly (item #50872175). Shown as (H) on image. The pre-trap is 50mm OD x 205mm in length and has two 35/20 socket joints to connect it to the manifold and main trap.
Catalog No. AF045402 Supplier Chemglass Life Sciences Supplier No. AF045402
Pre-Trap Only, Airfree, Schlenk. Pre-trap component of 4-Port Vacuum Manifold Right Handed Trap Assembly (item #50872175). Shown as (H) on image. The pre-trap is 50mm OD x 205mm in length and has two 35/20 socket joints to connect it to the manifold and main trap.