Learn More
Chemglass Life Sciences Pre-Trap Only (B), Airfree, Schlenk
SDP
Catalog No. AF020303
Encompass_Preferred
Pre-Trap Only (B), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. Pre-trap (B) is 51mm OD x 250mm long with 28/15 polished socket joints to connect the line shut off adapter (A) to the main trap stopper assembly (C) which has 28/15 o-ring ball joints. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds. Complete item is supplied with one #331 Viton o-ring, one CG-139-02 horseshoe clamp, three #116 Viton o-rings and two CG-150-05 #28 Stainless Steel pinch clamps.
Catalog No. AF020303 Supplier Chemglass Life Sciences Supplier No. AF020303
Pre-Trap Only (B), Airfree, Schlenk. Dual trap assembly for use with AF-0060 or AF-0061 manifolds. Pre-trap (B) is 51mm OD x 250mm long with 28/15 polished socket joints to connect the line shut off adapter (A) to the main trap stopper assembly (C) which has 28/15 o-ring ball joints. Trap assembly is designed to allow connection to either the right or left side of AF-0060 or AF-0061 manifolds. Complete item is supplied with one #331 Viton o-ring, one CG-139-02 horseshoe clamp, three #116 Viton o-rings and two CG-150-05 #28 Stainless Steel pinch clamps.