Learn More
Valutek™ Pre-Wetted Polyester Wiper, Cold Knife Cut
Catalog No. 191502034
Change view
Click to view available options
Packaging Quantity:
150/Pail
75/refill Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-150-2034 75/refill Bag
19-150-2033 150/Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-2034 Supplier Valutek™ Supplier No. VTPNWSI70R99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Composed of 100% continuous-filament polyester fabric pre-wetted with a blend of isopropyl alcohol and DI water.
  • The pre-wetted format enhances the wiper’s absorbent capacity, effectiveness, and reduces airborne contamination from traditional solvent bottle dispensers, spray and spills.
  • 100% continuous filament, double-knit polyester fiber with cold knife cut edge
  • Pre-wetted with specified IPA/DI Water concentrations to a 38% saturation level
  • Available in both reusable pail packaging and refill pack packaging
    The pre-wet solution increases both cleaning efficiency and cleaning protocol consistency
  • Very low levels of particles, fibers, ions and extractables
  • High abrasion resistance
  • All Valutek™ pre-wetted wipers are assembled in a Class 10 cleanroom under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 1-10 (ISO 3-4) critical environment
  • It is designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required
  • Various IPA blend levels are ideal for removing different cleaning and disinfecting residues in regulated environments
Specifications
Material Polyester
Dimensions (L x W) 9 x 9 in.
Packaging Quantity 75/refill Bag
Sterility Non-sterile