Learn More
Miles Scientific RP18 PREADSORBENT 200UM 20X20CM (25 PLATES/BOX)
SDP
Catalog No. NC0684933
Encompass_Preferred
Reversed Phase Octadecyl Silanized Silica Gel. These reversed phase TLC plates have been manufacturer to be greater than 50% and less than 100% silanized. Because of this they are hydrophobic, but wettable (unlike CAT# 164077 & 165077 which are 100% silanized). FEATURES: - 14% carbon content - Acid stable (UV254) fluorescent indicator - 20x20 cm glass backing - 250um coating thickness - 25 plates/box - Recomended Applications: aminophenols, barbiturates, preservatives, nucleobases, polycyclic aromatic hydrocarbons, steroids, tetracyclines, plasticizers (phthalates)
Catalog No. NC0684933 Supplier Miles Scientific Supplier No. P90011
Reversed Phase Octadecyl Silanized Silica Gel. These reversed phase TLC plates have been manufacturer to be greater than 50% and less than 100% silanized. Because of this they are hydrophobic, but wettable (unlike CAT# 164077 & 165077 which are 100% silanized). FEATURES: - 14% carbon content - Acid stable (UV254) fluorescent indicator - 20x20 cm glass backing - 250um coating thickness - 25 plates/box - Recomended Applications: aminophenols, barbiturates, preservatives, nucleobases, polycyclic aromatic hydrocarbons, steroids, tetracyclines, plasticizers (phthalates)