Learn More
Aat Bioquest ReadiUse™ Preactivated HRP NHS ester
SDP
Catalog No. 501953335
Encompass_Preferred
ReadiUse™ Preactivated HRP NHS ester is the mono NHS ester of HRP. It can be used to readily label proteins (such as antibodies) or other biologicals molecules that have an amino group.
Catalog No. 50-195-3335 Supplier Aat Bioquest Supplier No. 11025
ReadiUse™ Preactivated HRP NHS ester is the mono NHS ester of HRP. It can be used to readily label proteins (such as antibodies) or other biologicals molecules that have an amino group.