Learn More
Promocell GmbH PromoCell Preadipocyte Differentiation Medium KIT 500 ml, Contains: C-27411, C-39437
Catalog No. 50306274
Encompass
The Preadipocyte Differentiation Medium is a defined and animal-component free medium, which induces the differentiation of preadipocytes into mature adipocytes. The mature adipocytes can be cultivated using the Adipocyte Nutrition Medium.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Differentiation Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Differentiation Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml || IBMX | 44 μg / ml || L-Thyroxine | 9 ng / ml || Ciglitazone | 3 μg / ml |
Catalog No. 50-306-274 Supplier Promocell GmbH Supplier No. C27437
The Preadipocyte Differentiation Medium is a defined and animal-component free medium, which induces the differentiation of preadipocytes into mature adipocytes. The mature adipocytes can be cultivated using the Adipocyte Nutrition Medium.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Differentiation Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Differentiation Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration || d-Biotin | 8 μg / ml || Insulin (recombinant human) | 0.5 μg / ml || Dexamethasone | 400 ng / ml || IBMX | 44 μg / ml || L-Thyroxine | 9 ng / ml || Ciglitazone | 3 μg / ml |