Learn More
Promocell GmbH PromoCell Preadipocyte Growth Medium 500 ml, ready-to-use, low serum, with C-39425 attached to the flask
Supplier:  Promocell GmbH C27410
Encompass
PromoCell Preadipocyte Growth Medium is a low-serum (5% V/V) medium developed for the undifferentiated expansion of human preadipocytes.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine preadipocytes/adipocytes.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* Growth Medium Kit: Includes Basal Medium and SupplementPack* SupplementMix: Contains all media supplements premixed in one vial* SupplementPack: Contains all media supplements as individual vials| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Endothelial Cell Growth Supplement | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Hydrocortisone | 1 μg / ml || Heparin | 90 μg / ml |
Catalog No. 50-306-267