PGM -2 Preadipocyte Growth Medium-2 BulletKit provides both a proliferation medium and a differentiation medium guaranteed to induce differentiation into mature, functionally active adipocytes.Each bottle of the PGM-2 BulletKit Medium contains enough differentiation medium to fully differentiate 10 million preadipocytes [10 complete 96-well plates] into mature, functionally active adipocytes.The PGM-2 Medium is offered as a BulletKit Medium (catalog no. PT-8002) which includes both the basal medium and the necessary supplements for both proliferation and differentiation of preadipocytes.The basal medium (PGM-2 Basal Medium; catalog no. PT-8202) and the necessary supplements (PGM-2 SingleQuot Kit; catalog no. PT-9502) can also be purchased individually.Content:1 x PGM-2 Basal Medium (PT-8202)1 x PGM-2 SingleQuots Supplement and Growth Factor Pack (PT-9502)