Learn More
Aviva Systems Biology Corp PREALBUMIN ELISA KIT MOUSE, 96 Wells, Sandwich ELISA, Detection Range: 15.6 ng/ml - 500 ng/ml, Sensitivity: 3.967 ng/ml, Colorimetric Detection, Suitable for Serum and Plasma
SDP
Supplier:  Aviva Systems Biology Corp OKIA00111
Encompass
PREALBUMIN ELISA KIT MOUSE, 96 Wells, Sandwich ELISA, Detection Range: 15.6 ng/ml - 500 ng/ml, Sensitivity: 3.967 ng/ml, Colorimetric Detection, Suitable for Serum and Plasma
Catalog No. NC1841092