Learn More
Crystal Chem Inc Prealbumin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80732
Encompass
The Prealbumin ELISA is used to quantify levels of human prealbumin in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 3.13 ng/mL. Dynamic range of 3.13 - 100 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7850