Learn More
Alpco Ltd Rat Pre-Albumin ELISA
SDP
Supplier:  Alpco Ltd 41PALRTE01
Encompass
The Rat Prealbumin ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Prealbumin in biological samples of rats. Prealbumin is a tetrameric protein consisting of four identical polypeptide chains. It migrates more rapidly than serum albumin on electrophoresis. It can be used as a marker in the nutritional evaluation of patients with a variety of diseases.
Catalog No. 50-751-3606