Learn More
Chemglass Life Sciences Preassembled SMOOTH Closure and Septa, 9mm Thread, Green, PTFE/Red Rubber Septa
SDP
Catalog No. 50231077
Encompass_Preferred
Preassembled SMOOTH Closure and Septa, 9mm Thread, Green, PTFE/Red Rubber Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-231-077 Supplier Chemglass Life Sciences Supplier No. CV3790G009
Preassembled SMOOTH Closure and Septa, 9mm Thread, Green, PTFE/Red Rubber Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.