Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Blue, 9mm Thread, PTFE/Silicone Septa
SDP
Catalog No. 50231082
Encompass_Preferred
Preassembled Closure and Septa, Blue, 9mm Thread, PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-231-082 Supplier Chemglass Life Sciences Supplier No. CV3800B009
Preassembled Closure and Septa, Blue, 9mm Thread, PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.