Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Red, 9mm Thread, PTFE/Silicone Septa
SDP
Catalog No. 50231085
Encompass_Preferred
Preassembled Closure and Septa, Red, 9mm Thread, PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-231-085 Supplier Chemglass Life Sciences Supplier No. CV3800R009
Preassembled Closure and Septa, Red, 9mm Thread, PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.