Learn More
Perkin Elmer US LLC Pre-Assembled Magnetic Steel Screw Cap with PTFE/Silicone (Blue/White) Septa - 0.06 in thick, 100/pk
Catalog No. 502104568
Encompass_Preferred
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
Catalog No. 50-210-4568 Supplier Perkin Elmer US LLC Supplier No. N6356476
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.