Learn More
Bio Rad Laboratories Precision Plus Protein Dual Color Standards, 2.5 mL
Supplier:  Bio Rad Laboratories 1610394
Encompass
2.5 mL (5 x 500 µl), mixture of 10 recombinant proteins (10–250 kD), 8 blue-stained bands, and 2 pink reference bands (25 and 75 kD), 250 applications.
Catalog No. NC1085839