Learn More
BIOFARGO INC Precast Agarose Gel, 1.5% agarose, with GelRed¬Æ Nucleic Acid Gel Stain (10000X), with TAE (11 well, DNA Size Marker 100bp - 2kp)
Catalog No. NC3880889
Encompass_Preferred
The Precast Agarose Gel Agarose  with GelRed Nucleic Acid Gel Stain from Biofargo is a ready‑to‑use electrophoresis kit featuring a 1-2% agarose gel cast in TAE buffer and pre‑stained with GelRed, saving you time on gel preparation and staining. The kit includes 6-50 wells per gel, optimized loading buffer, and DNA markers for streamlined nucleic acid separation workflows. With GelRed offering superior sensitivity and safety compared to traditional ethidium bromide, this product is ideal for routine PCR checks and fragment analysis where fast setup and consistent results matter.
Catalog No. NC3880889 Supplier BIOFARGO INC Supplier No. B10F1511OM200015
The Precast Agarose Gel Agarose  with GelRed Nucleic Acid Gel Stain from Biofargo is a ready‑to‑use electrophoresis kit featuring a 1-2% agarose gel cast in TAE buffer and pre‑stained with GelRed, saving you time on gel preparation and staining. The kit includes 6-50 wells per gel, optimized loading buffer, and DNA markers for streamlined nucleic acid separation workflows. With GelRed offering superior sensitivity and safety compared to traditional ethidium bromide, this product is ideal for routine PCR checks and fragment analysis where fast setup and consistent results matter.