Learn More
Sigma Aldrich Fine Chemicals Biosciences Precast Agarose Gels 8 wel
Catalog No. 502904744
Encompass
Precast agarose gels are designed for high reSOLUTIONon separation of nucleic acids producing sharp RNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays and are supplied in a box of twenty.Gel size 6.0 x 9.5 cm 5.5 mm thickCassette size 6.8 x 10.2 cm container tray (fits common horizontal electrophoresis units)Well orientation 8-well portrait style
Catalog No. 50-290-4744 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P62221PAK
Precast agarose gels are designed for high reSOLUTIONon separation of nucleic acids producing sharp RNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays and are supplied in a box of twenty.Gel size 6.0 x 9.5 cm 5.5 mm thickCassette size 6.8 x 10.2 cm container tray (fits common horizontal electrophoresis units)Well orientation 8-well portrait style