Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Precast Gel Plus, Tris-Gly 12%

Catalog No. p-7251025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Wells:
10-well
15-well
Sample Loading Volume:
≤30 μL
≤50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Sample Loading Volume
ICN08W00003 10-well ≤50 μL
ICN08W00004 15-well ≤30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN08W00003 Supplier MP Biomedicals™ Supplier No. 08W00003
Only null left
Add to Cart
Add to Cart

These precast polyacrylamide gels are suitable for both SDS-PAGE and Native-PAGE electrophoresis followed by gel staining or Western blotting.

Precast Gel Plus gels are an efficient and convenient scientific tool for separating peptides and small proteins with clear band resolution. In addition, they are compatible with Bio-Rad Mini Protean Tanks, thus facilitating the use of our gels without the need to replace your current hardware.

  • Ready-to-use gels for SDS-PAGE or Native-PAGE applications
  • Resilient to mechanical stretching during handling
  • Removable comb prevents well-wall breakage
  • Outlined and numbered wells facilitate gel loading and tracking
  • Easy to store and stable at room temperature for 6 months
  • Do not freeze
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C for 12 Months
Buffer MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE
Color Clear
Concentration 12%
Gel Percentage 12%
Gel Size Mini
Gel Thickness 1 mm
Gel Type Tris-Glycine
Sample Loading Volume ≤50 μL
Separation of Peptides, Proteins
Separation Range 15 to 150 kDa
Separation Type Denaturing, Native
Wells 10-well
For Use With (Application) Gel Staining SDS-PAGE, Native-PAGE, Blue Native, Western Blot, Acrylamide, Tris-Glycine, Mini Gels, Electrophoresis
Quantity 10 Cassettes
Product Type Precast Gel
For Use With (Equipment) Bio-Rad Mini Protean Tanks
Show More Show Less

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.