Learn More
Electron Microscopy Sciences T-Sue™ Precast Paraffin Block, 2 mm
SDP
Supplier:  Electron Microscopy Sciences 6913302
Encompass_Preferred
Precast T-Sue Microarray paraffin blocks are used to construct a tissue array block in minutes by simply punching the donor tissue cores and inserting into a pre-made paraffin recipient block. It is an easy, convenient, and low cost method for constructing paraffin tissue microarrays. The blocks are made of paraffin and the cassettes are made of acetal. The cassette is permanently bonded to the block and has consistent core diameters. It is ready to accept cores from the donor block.
Kit includes 6 Precast T-Sue Paraffin blocks and two punch needles with stylet.
Size: 2 mm
Capacity: 24 cores
Catalog No. 50-193-0040