Learn More
Bio Rad Laboratories 10% Mini-PROTEAN TGX Precast Protein Gels, 10-well, 50 uL
Catalog No. NC1172307
Encompass
Use these Mini-PROTEAN TGX Precast Protein Gels for separation of polypeptides ~20–200 kD. Long-life TGX (Tris-Glycine eXtended) Gels have a novel formulation and can be used for both standard denaturing protein separations as well as native electrophoresis.
Catalog No. NC1172307 Supplier Bio Rad Laboratories Supplier No. 4561034
Use these Mini-PROTEAN TGX Precast Protein Gels for separation of polypeptides ~20–200 kD. Long-life TGX (Tris-Glycine eXtended) Gels have a novel formulation and can be used for both standard denaturing protein separations as well as native electrophoresis.