Learn More
Cayman Chemical Precellys 0 5 mL Soft Tissue
SDP
Catalog No. 502886483
Encompass_Preferred
Kit includes 50 preps of 1.4 mm ceramic (zirconium oxide) beads in 0.5 mL standard tubes. CK14 0.5 mL is designed for soft tissue homogenization like brain, liver, kidney, artery, and more. Sample size range from 1 mg to 20 mg tissue or 5 µL to 200 µL culture. [Bertin Catalog P000933-LYSK0-A.0; Previously KT03961-1-203.05]
Catalog No. 50-288-6483 Supplier Cayman Chemical Supplier No. 1042810 BOX
Kit includes 50 preps of 1.4 mm ceramic (zirconium oxide) beads in 0.5 mL standard tubes. CK14 0.5 mL is designed for soft tissue homogenization like brain, liver, kidney, artery, and more. Sample size range from 1 mg to 20 mg tissue or 5 µL to 200 µL culture. [Bertin Catalog P000933-LYSK0-A.0; Previously KT03961-1-203.05]