Learn More
Cayman Chemical Precellys 0 5 mL Soft Tissue
SDP
Supplier:  Cayman Chemical 104281 BOX
Encompass_Preferred
Kit includes 50 preps of 1.4 mm ceramic (zirconium oxide) beads in 0.5 mL standard tubes. CK14 0.5 mL is designed for soft tissue homogenization like brain, liver, kidney, artery, and more. Sample size range from 1 mg to 20 mg tissue or 5 µL to 200 µL culture. [Bertin Catalog P000933-LYSK0-A.0; Previously KT03961-1-203.05]
Catalog No. 50-288-7109