Learn More
Cayman Chemical Precellys 0 5 mL Tough Micro
SDP
Catalog No. 502886500
Encompass_Preferred
Kit includes 50 preps of 0.5 mm glass beads in 0.5 mL standard tubes. VK05 0.5 mL is designed for tough micro-organism lysing like yeast, fungi and more. Sample size range from 1 mg to 20 mg sample or 5 µL to 200 µL culture. [Bertin Catalog P000934-LYSK0-A.0; Previously KT03961-1-204.05]
Catalog No. 50-288-6500 Supplier Cayman Chemical Supplier No. 104302 BOX
Kit includes 50 preps of 0.5 mm glass beads in 0.5 mL standard tubes. VK05 0.5 mL is designed for tough micro-organism lysing like yeast, fungi and more. Sample size range from 1 mg to 20 mg sample or 5 µL to 200 µL culture. [Bertin Catalog P000934-LYSK0-A.0; Previously KT03961-1-204.05]