Learn More
Cayman Chemical Precellys 15 mL Bacteria Lys
SDP
Supplier:  Cayman Chemical 169131 BOX
Encompass_Preferred
Precellys Bacteria Lysing Kit includes 25 preps of 0.1 mm ceramic beads (CK01) in 15 ml tubes. CK01 beads are designed for micro-organism lysing like gram+, gram-bacteria, spores and more. Sample size range from 400 mg to 4 g sample or 400 µl to 9 ml culture. [Bertin Catalog P000948-LYSK0-A.0; Previously KT03961-1-510.15]
Catalog No. 50-296-5221