Learn More
Cayman Chemical Precellys 15 mL Bacteria Lys
SDP
Catalog No. 502965192
Encompass_Preferred
Precellys Bacteria Lysing Kit includes 25 preps of 0.1 mm ceramic beads (CK01) in 15 ml tubes. CK01 beads are designed for micro-organism lysing like gram+, gram-bacteria, spores and more. Sample size range from 400 mg to 4 g sample or 400 µl to 9 ml culture. [Bertin Catalog P000948-LYSK0-A.0; Previously KT03961-1-510.15]
Catalog No. 50-296-5192 Supplier Cayman Chemical Supplier No. 169132 BOX
Precellys Bacteria Lysing Kit includes 25 preps of 0.1 mm ceramic beads (CK01) in 15 ml tubes. CK01 beads are designed for micro-organism lysing like gram+, gram-bacteria, spores and more. Sample size range from 400 mg to 4 g sample or 400 µl to 9 ml culture. [Bertin Catalog P000948-LYSK0-A.0; Previously KT03961-1-510.15]