Learn More
Cayman Chemical Precellys 2 mL Bacteria Lysi
SDP
Supplier:  Cayman Chemical 139332 BOX
Encompass_Preferred
Kit includes 50 preps of 0.1 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK01 is designed for micro-organism lysing like, gram+, gram- bacteria, spores and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000919-LYSK0-A.0; Previously KT03961-1-010.2]
Catalog No. 50-295-4291