Learn More
Cayman Chemical Precellys 2 mL Dry Hard Tiss
SDP
Supplier:  Cayman Chemical 152352 BOX
Encompass_Preferred
Kit includes 50 preps of ceramic (zirconium oxide) beads of 6.8 mm in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. CK68 Reinforced is designed for dry samples grinding like rice grains, pepper grains and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000923-LYSK0-A.0; Previously KT03961-1-014.2]
Catalog No. 50-297-2821