Learn More
Cayman Chemical Precellys 2 mL Empty Tubes a
SDP
Catalog No. 502971613
Encompass_Preferred
Standard 2 mL empty tubes suitable for use with the Precellys Evolution Touch (38707), Precellys 24, and Precellys Minilys Personal Tissue Homogenizer (Item No. 10435). (Qty: 500 tubes/box) [Bertin Catalog P000945-LYSK0-A.0; Previously KT03961-1-405.2]
Catalog No. 50-297-1613 Supplier Cayman Chemical Supplier No. 152361 BOX
Standard 2 mL empty tubes suitable for use with the Precellys Evolution Touch (38707), Precellys 24, and Precellys Minilys Personal Tissue Homogenizer (Item No. 10435). (Qty: 500 tubes/box) [Bertin Catalog P000945-LYSK0-A.0; Previously KT03961-1-405.2]