Learn More
Cayman Chemical Precellys 2 mL Hard Tissue H
SDP
Catalog No. 502886833
Encompass_Preferred
Kit includes 50 preps of 2.8 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK28 beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000911-LYSK0-A.0; Previously KT03961-1-002.2]
Catalog No. 50-288-6833 Supplier Cayman Chemical Supplier No. 100111512 BOX
Kit includes 50 preps of 2.8 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK28 beads are designed for hard tissue homogenization like EAr, lung, heart, spleen, tail, artery, arabidopsis, leaves, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000911-LYSK0-A.0; Previously KT03961-1-002.2]