Learn More
Cayman Chemical Precellys 2 mL Hard Tissue R
SDP
Supplier:  Cayman Chemical 168581 BOX
Encompass_Preferred
Kit includes 50 preps of 2.8 mm stainless steel beads in 2 mL reinforced tubes. Custom design reinforced tubes for bead beating application: O-ring and reinforced materials for dry grinding, valuable or very hard samples. MK28-R is designed for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, corn, rice and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000917-LYSK0-A.0; Previously KT03961-1-008.2]
Catalog No. 50-296-5704