Learn More
Cayman Chemical Precellys 2 mL Microorganism
SDP
Supplier:  Cayman Chemical 100111552 BOX
Encompass_Preferred
Kit includes 50 preps of 0.1 mm glass beads in 2 mL standard tubes. VK01 is designed for micro-organism lysing like gram-, gram+ bacteria, spores and more. Sample size range from 20 mg to 200 mg sample or 200 µL to 1.6 mL culture. [Bertin Catalog P000914-LYSK0-A.0; Previously KT03961-1-005.2]
Catalog No. 50-288-6928